Nickel powder, spherical, -100+325 mesh, 99.8% (metals basis), Thermo Scientific™
Nickel powder, spherical, -100+325 mesh, 99.8% (metals basis), Thermo Scientific™
Nickel powder, spherical, -100+325 mesh, 99.8% (metals basis), Thermo Scientific™
Thermo Scientific™

Nickel powder, spherical, -100+325 mesh, 99.8% (metals basis), Thermo Scientific™

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number 042733.30
Price (USD)
-
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS7440-02-0
IUPAC Namenickel
Molecular FormulaNi
InChI KeyPXHVJJICTQNCMI-UHFFFAOYSA-N
SMILES[Ni]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.2% max.
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only