Potassium hydroxide, 0.1N Standardized Solution in methanol
Potassium hydroxide, 0.1N Standardized Solution in methanol
Potassium hydroxide, 0.1N Standardized Solution in methanol
Thermo Scientific Chemicals

Potassium hydroxide, 0.1N Standardized Solution in methanol

CAS: 1310-58-3 | HKO | 56.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 L
20 L
Catalog number 042734.K7
also known as 042734-K7
Price (USD)
113.00
Each
Quantity:
4 L
Request bulk or custom format
Price (USD)
113.00
Each
Chemical Identifiers
CAS1310-58-3
IUPAC Namepotassium hydroxide
Molecular FormulaHKO
InChI KeyKWYUFKZDYYNOTN-UHFFFAOYSA-M
SMILES[OH-].[K+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Concentration0.1N (0.14% uncertainty)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only