Polystyrene latex microsphere, 2 micron, 2.5 wt% dispersion in water
Polystyrene latex microsphere, 2 micron, 2.5 wt% dispersion in water
Polystyrene latex microsphere, 2 micron, 2.5 wt% dispersion in water
Thermo Scientific Chemicals

Polystyrene latex microsphere, 2 micron, 2.5 wt% dispersion in water

CAS: 9003-53-6 | (C8H8)n | 104.15 g/mol
Have Questions?
Catalog NumberQuantity
042744.4Y
also known as 042744-4Y
15 mL
Catalog number 042744.4Y
also known as 042744-4Y
Price (USD)
460.65
Online Exclusive
512.00
Save 51.35 (10%)
Each
Quantity:
15 mL
Request bulk or custom format
Price (USD)
460.65
Online Exclusive
512.00
Save 51.35 (10%)
Each
Chemical Identifiers
CAS9003-53-6
Molecular Formula(C8H8)n
SMILES*-CC(-*)C1=CC=CC=C1
MDL NumberMFCD00084450
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)1.80 to 2.20 micron in diamter
CommentCoefficient of variance: 0 - 5%
Assay (unspecified)2.5 to 2.8% solids content
Polystyrene latex microsphere is used as a biocompatible material, especially in dentistry. They are used as insulators, for injection molding and casting, as sheets, plates, rods, rigid forms and beads. It is also used as extrusion molded foamed packaging, dinnerware, bottles, toys and disposable cutlery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only