Polystyrene latex microsphere, 45.0 micron, 2.5 wt% dispersion in water
Polystyrene latex microsphere, 45.0 micron, 2.5 wt% dispersion in water
Polystyrene latex microsphere, 45.0 micron, 2.5 wt% dispersion in water
Thermo Scientific Chemicals

Polystyrene latex microsphere, 45.0 micron, 2.5 wt% dispersion in water

CAS: 9003-53-6 | (C8H8)n | 104.15 g/mol
Have Questions?
Catalog NumberQuantity
042749.AA
also known as 042749-AA
5 mL
Catalog number 042749.AA
also known as 042749-AA
Price (USD)
522.65
Special offer
Online exclusive
Ends: 30-Jun-2026
615.00
Save 92.35 (15%)
Each
Quantity:
5 mL
Request bulk or custom format
Price (USD)
522.65
Special offer
Online exclusive
Ends: 30-Jun-2026
615.00
Save 92.35 (15%)
Each
Chemical Identifiers
CAS9003-53-6
Molecular Formula(C8H8)n
SMILES*-CC(-*)C1=CC=CC=C1
MDL NumberMFCD00084450
SpecificationsSpecification SheetSpecification Sheet
FormDispersion
Assay (special)Solids content: 2.5-2.8%
Particle SizeDiameter: 40.0-50.0 micron
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only