Cresol Red sodium salt, 0.04% w/v aq. soln.
Cresol Red sodium salt, 0.04% w/v aq. soln.
Cresol Red sodium salt, 0.04% w/v aq. soln.
Cresol Red sodium salt, 0.04% w/v aq. soln.
Thermo Scientific Chemicals

Cresol Red sodium salt, 0.04% w/v aq. soln.

CAS: 62625-29-0 | C21H17O5S
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 042752.AP
also known as 042752-AP
Price (USD)
36.80
Each
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)
36.80
Each
Add to cart
Chemical Identifiers
CAS62625-29-0
IUPAC Namesodium 4-[3-(4-hydroxy-3-methylphenyl)-1,1-dioxo-3H-2,1λ⁶-benzoxathiol-3-yl]-2-methylbenzen-1-olate
Molecular FormulaC21H17NaO5S
InChI KeyFMKJVYROEYMYFV-UHFFFAOYNA-M
SMILES[Na+].CC1=CC(=CC=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C([O-])C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Transition IntervalTransition interval: pH 0.2 (red) to pH 1.8 (yellow); pH 7.2 (yellow) to pH 8.8 (red)
Concentration (w/v)Cresol Red sodium salt, 0.04% w/v aq. soln.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only