Methyl Purple, 0.1% w/v aq. soln.
Methyl Purple, 0.1% w/v aq. soln.
Methyl Purple, 0.1% w/v aq. soln.
Thermo Scientific Chemicals

Methyl Purple, 0.1% w/v aq. soln.

Transition interval: pH 4.8 (purple) to pH 5.4 (green) | CAS: 1340-02-9
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
Catalog number 042753.K2
also known as 042753-K2
Price (USD)
67.90
Each
Quantity:
1 L
Request bulk or custom format
Price (USD)
67.90
Each
Specifications
Assay Percent Range0.1% w/v aqueous solution
CAS1340-02-9
CAS7732-18-5
Chemical Name or MaterialMethyl Purple
Concentration or Composition (by Analyte or Components)Methyl purple: 0.1%; Water: 99.9%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only