Tetrakis(dimethylamino)germanium(IV), 99% (metals basis), Thermo Scientific Chemicals
Tetrakis(dimethylamino)germanium(IV), 99% (metals basis), Thermo Scientific Chemicals
Tetrakis(dimethylamino)germanium(IV), 99% (metals basis), Thermo Scientific Chemicals
Tetrakis(dimethylamino)germanium(IV), 99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetrakis(dimethylamino)germanium(IV), 99% (metals basis), Thermo Scientific Chemicals

Preparation of nitrides
Have Questions?
Catalog NumberQuantity
042760.09
also known as 042760-09
10 g
Catalog number 042760.09
also known as 042760-09
Price (USD)
-
Quantity:
10 g
Request bulk or custom format
Specifications
Assay Percent Notes(metals basis)
CAS7344-40-3
Chemical Name or MaterialTetrakis (dimethylamino)germanium (IV)
ColorColorless
DOT InformationTransport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: WATER-REACTIVE LIQUID, CORROSIVE, N.O.S.
View more
Preparation of nitrides

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only