Silica gel, TLC high purity grade, with gypsum binder and fluorescent indicator
Silica gel, TLC high purity grade, with gypsum binder and fluorescent indicator
Silica gel, TLC high purity grade, with gypsum binder and fluorescent indicator
Thermo Scientific Chemicals

Silica gel, TLC high purity grade, with gypsum binder and fluorescent indicator

TLC plate preparation | CAS: 63231-67-4 | H2O3Si | 78.098 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number 042764.30
also known as 042764-30
Price (USD)
109.65
Special offer
Online exclusive
Ends: 28-Dec-2026
129.00
Save 19.35
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
109.65
Special offer
Online exclusive
Ends: 28-Dec-2026
129.00
Save 19.35
Each
Specifications
CAS63231-67-4
Chemical Name or MaterialSilica gel
EINECS Number231-545-4
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H373
May cause damage to organs through prolonged or repeated exposure.
View more
Silica gel is primarily used in plate preparation of thin layer chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only