Methylene Blue, 1% w/v aq. soln.
Methylene Blue, 1% w/v aq. soln.
Methylene Blue, 1% w/v aq. soln.
Methylene Blue, 1% w/v aq. soln.
Thermo Scientific Chemicals

Methylene Blue, 1% w/v aq. soln.

CAS: 122965-43-9 | C16H18ClN3S
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 042771.AE
also known as 042771-AE
Price (USD)
35.10
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
35.10
Each
Specifications
Assay Percent Range1% w/v aqueous solution
CAS122965-43-9
CAS7732-18-5
Chemical Name or MaterialMethylene Blue
Concentration or Composition (by Analyte or Components)3, 7-Bis(dimethylamino)phenothiazin-5-ium chloride hydrate: 1.0%; Water: 99%
View more

Methylene Blue, 1% w/v aqueous solution is used as a dye in different staining procedures viz. Wright's stain and Jenner's stain. It also serves as an indicator and medicine. Further, it is used to examine RNA and DNA under the microscope or in a gel. It is widely used as a redox indicator in analytical chemistry. It is also used in the sulfide analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only