Vanadium pieces, 99.7% (metals basis)
Vanadium pieces, 99.7% (metals basis)
Vanadium pieces, 99.7% (metals basis)
Thermo Scientific Chemicals

Vanadium pieces, 99.7% (metals basis)

CAS: 7440-62-2 | V | 50.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number 042775.30
also known as 042775-30
Price (USD)
731.65
Online Exclusive
813.00
Save 81.35 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
731.65
Online Exclusive
813.00
Save 81.35 (10%)
Each
Specifications
AppearanceSilver Gray pieces
Assay Percent Range99.7% (Metals basis)
CAS7440-62-2
ChEBICHEBI:35170
Chemical Name or MaterialVanadium
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only