Dimethyl sulfoxide, HPLC grade, 99.9+%, packaged under Argon in resealable ChemSeal™ bottles
Dimethyl sulfoxide, HPLC grade, 99.9+%, packaged under Argon in resealable ChemSeal™ bottles
Dimethyl sulfoxide, HPLC grade, 99.9+%, packaged under Argon in resealable ChemSeal™ bottles
Thermo Scientific Chemicals

Dimethyl sulfoxide, HPLC grade, 99.9+%, packaged under Argon in resealable ChemSeal™ bottles

Sensitivity: Hygroscopic | CAS: 67-68-5 | C2H6OS | 78.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
4 x 1 L
Catalog number 042780.AK
also known as 042780-AK
Price (USD)
42.65
Special offer
Online exclusive
Ends: 30-Jun-2026
49.90
Save 7.25 (15%)
Each
Quantity:
250 mL
Request bulk or custom format
Price (USD)
42.65
Special offer
Online exclusive
Ends: 30-Jun-2026
49.90
Save 7.25 (15%)
Each
Chemical Identifiers
CAS67-68-5
IUPAC Namemethanesulfinylmethane
Molecular FormulaC2H6OS
InChI KeyIAZDPXIOMUYVGZ-UHFFFAOYSA-N
SMILESCS(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid. HPLC grade, packaged under Argon in resealable ChemSeal™ bottles
Optical Absorbance at275 nm ≤ 0.50 AU
Optical Absorbance at400 nm ≤ 0.005 AU
Water Content (Karl Fischer Titration)≤0.1 %
Optical Absorbance at350 nm ≤ 0.02 AU
View more
Dimethyl sulfoxide is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only