Thymol Blue sodium salt, ACS, Thermo Scientific Chemicals
Thymol Blue sodium salt, ACS, Thermo Scientific Chemicals
Thymol Blue sodium salt, ACS, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Thymol Blue sodium salt, ACS, Thermo Scientific Chemicals

Water soluble pH indicator
Have Questions?
Catalog NumberQuantity
042785.09
also known as 042785-09
10 g
Catalog number 042785.09
also known as 042785-09
Price (USD)
50.65
Online Exclusive
56.40
Save 5.75 (10%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
50.65
Online Exclusive
56.40
Save 5.75 (10%)
Each
Chemical Identifiers
CAS62625-21-2
IUPAC Namesodium 2-{[4-hydroxy-2-methyl-5-(propan-2-yl)phenyl][(1E)-2-methyl-4-oxo-5-(propan-2-yl)cyclohexa-2,5-dien-1-ylidene]methyl}benzene-1-sulfonate
Molecular FormulaC27H29NaO5S
InChI KeyOCMIKNBSIRSUPI-FRWNMSGJSA-M
SMILES[Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
CommentClarity of solution P.T.
CommentVisual transition interval (alkaline range) pH 8.0 (yellow) to pH 9.2 (blue)
CommentVisual transition interval (acid range) pH 1.2 (pink) to pH 2.8 (yellow)
Water soluble pH indicatorThymol Blue sodium salt acts as a pH indicator and utilized to indicate the changes in pH. It is involved in the ionization in multi-component drug formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only