Glassy carbon plate, 3mm (0.1in) thick, type 2
Glassy carbon plate, 3mm (0.1in) thick, type 2
Glassy carbon plate, 3mm (0.1in) thick, type 2
Thermo Scientific Chemicals

Glassy carbon plate, 3mm (0.1in) thick, type 2

Used in casting, smelting of metal, hard alloys. Glassy carbon is widely used as an electrode material in electrochemistry, for high temperature crucibles and as a part of some prosthetic devices. | CAS: 7440-44-0
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
Catalog number 042820.FI
also known as 042820-FI
Price (USD)
353.65
Online exclusive
393.00
Save 39.35
Each
Quantity:
50 x 50 mm
Request bulk or custom format
Price (USD)
353.65
Online exclusive
393.00
Save 39.35
Each
Chemical Identifiers
CAS7440-44-0
Specifications
CAS7440-44-0
Chemical Name or MaterialGlassy carbon plate
EINECS Number231-153-3
MDL NumberMFCD00133992
PackagingTube
View more
Used in casting, smelting of metal, hard alloys. Glassy carbon is widely used as an electrode material in electrochemistry, for high temperature crucibles and as a part of some prosthetic devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Frequently asked questions (FAQs)

What is the difference between type 1 glassy carbon and type 2 glassy carbon?

Type 1 is a "low temperature" glassy carbon produced by heat treatment at ~ 1,000-2,000°C and consists of discreet fragments of distorted graphene layers. In general, type 1 is less compressible, more resistant to oxidation, and less permeable to gasses. Type 2 is a"high temperature" glassy carbon produced by heat treatment at ~2,500-3,000°C and consists of broken or imperfect fullerene-like nanospheroids encased in a disordered, multilayered graphene matrix. Type 2 usually contains fewer impurities and has higher electrical and thermal conductivity.