Glassy carbon rod, 4mm (0.16in) dia, type 2
Glassy carbon rod, 4mm (0.16in) dia, type 2
Glassy carbon rod, 4mm (0.16in) dia, type 2
Thermo Scientific Chemicals

Glassy carbon rod, 4mm (0.16in) dia, type 2

CAS: 7440-44-0
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mm
100 mm
200 mm
Catalog number 042825.DM
also known as 042825-DM
Price (USD)
582.65
Online Exclusive
647.00
Save 64.35 (10%)
Each
Quantity:
100 mm
Request bulk or custom format
Price (USD)
582.65
Online Exclusive
647.00
Save 64.35 (10%)
Each
Chemical Identifiers
CAS7440-44-0
IUPAC Namecarbon
Molecular FormulaC
InChI KeyOKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES[C]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Black
FormRod, 4mm (0.16in) diameter (Glassy Carbon type-2, maximimum service temperature 3000°C).
Assay from Suppliers CofAReport element content
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Frequently asked questions (FAQs)

What is the difference between type 1 glassy carbon and type 2 glassy carbon?

Type 1 is a "low temperature" glassy carbon produced by heat treatment at ~ 1,000-2,000°C and consists of discreet fragments of distorted graphene layers. In general, type 1 is less compressible, more resistant to oxidation, and less permeable to gasses. Type 2 is a"high temperature" glassy carbon produced by heat treatment at ~2,500-3,000°C and consists of broken or imperfect fullerene-like nanospheroids encased in a disordered, multilayered graphene matrix. Type 2 usually contains fewer impurities and has higher electrical and thermal conductivity.