Antimony(III) iodide, ultra dry, 99.999% (metals basis)
Antimony(III) iodide, ultra dry, 99.999% (metals basis)
Antimony(III) iodide, ultra dry, 99.999% (metals basis)
Thermo Scientific Chemicals

Antimony(III) iodide, ultra dry, 99.999% (metals basis)

CAS: 7790-44-5 | I3Sb | 502.47 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
Catalog number 042831.09
also known as 042831-09
Price (USD)
230.65
Online Exclusive
256.00
Save 25.35 (10%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
230.65
Online Exclusive
256.00
Save 25.35 (10%)
Each
Chemical Identifiers
CAS7790-44-5
IUPAC Nameantimony(3+) triiodide
Molecular FormulaI3Sb
InChI KeyKWQLUUQBTAXYCB-UHFFFAOYSA-K
SMILES[Sb+3].[I-].[I-].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)brown to very dark brown and red to very dark red and red-brown
Formpowder or crystals or granules or chunks
Residual water< 100 ppm
Total Metal Impurities0.001% max.
Antimony(III) iodide is used as a dopant in the preparation of thermoelectric materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only