Cesium chloride, ultra dry, 99.9% (metals basis)
Cesium chloride, ultra dry, 99.9% (metals basis)
Cesium chloride, ultra dry, 99.9% (metals basis)
Thermo Scientific Chemicals

Cesium chloride, ultra dry, 99.9% (metals basis)

CAS: 7647-17-8 | ClCs | 168.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 042832.22
also known as 042832-22
Price (USD)
392.65
Special offer
Online exclusive
Ends: 30-Jun-2026
462.00
Save 69.35 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
392.65
Special offer
Online exclusive
Ends: 30-Jun-2026
462.00
Save 69.35 (15%)
Each
Chemical Identifiers
CAS7647-17-8
IUPAC Namecaesium(1+) chloride
Molecular FormulaClCs
InChI KeyAIYUHDOJVYHVIT-UHFFFAOYSA-M
SMILES[Cl-].[Cs+]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White
Total Metal Impurities0.1% max.
Residual water< 100 ppm
Cesium chloride is widely used in isopycnic centrifugation for separating various types of DNA. It is a reagent in analytical chemistry, where it is used to identify ions by the color and morphology of the precipitate. When enriched in radioisotopes, such as 137CsCl or 131CsCl, cesium chloride is used in nuclear medicine applications such as treatment of cancer and diagnosis of myocardial infarction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only