Nickel rod, 5mm (0.2in) dia, Puratronic™, 99.999% (metals basis) - FAQs
View additional product information for Nickel rod, 5mm (0.2in) dia, Puratronic™, 99.999% (metals basis) - FAQs (042835.DJ, 042835.DO)
2 product FAQs found
What is the weight of the 150 mm size Nickel rod, 5mm (0.2in) dia, Puratronic, 99.999% (metals basis)?
The weight of the 150 mm nickel rod is ≈26.25 g.
What is the weight of the 50 mm size Nickel rod, 5mm (0.2in) dia, Puratronic, 99.999% (metals basis)?
The weight of the 50 mm nickel rod is ≈8.75 g.