Manganese(II) chloride, ultra dry, 99.99% (metals basis)
Manganese(II) chloride, ultra dry, 99.99% (metals basis)
Manganese(II) chloride, ultra dry, 99.99% (metals basis)
Thermo Scientific Chemicals

Manganese(II) chloride, ultra dry, 99.99% (metals basis)

Plays an important role in the production of dry cell batteries. | CAS: 7773-01-5 | Cl2Mn | 125.84 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
Catalog number 042844.18
also known as 042844-18
Price (USD)
370.65
Online Exclusive
412.00
Save 41.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
370.65
Online Exclusive
412.00
Save 41.35 (10%)
Each
Chemical Identifiers
CAS7773-01-5
IUPAC Namemanganese(2+) dichloride
Molecular FormulaCl2Mn
InChI KeyGLFNIEUTAYBVOC-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Mn++]
View more
SpecificationsSpecification SheetSpecification Sheet
Residual water< 100 ppm
Total Metal Impurities0.01% max.
FormPowder
Appearance (Color)Pink
Manganese(II) chloride, plays an important role in the production of dry cell batteries. It is also used as a starting material to the methylcyclopentadienyl manganese tricarbonyl, which has antiknock property. Further, it is used in the formation of an active Manganese(0) species useful for radical cyclization. It is also used in 31P-NMR to determine the size and lamellarity of phospholipidvesicles reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only