Niobium slug, 3.175mm (0.125in) dia x 3.175mm (0.125in) length, 99.95% (metals basis excluding Ta)
Niobium slug, 3.175mm (0.125in) dia x 3.175mm (0.125in) length, 99.95% (metals basis excluding Ta)
Niobium slug, 3.175mm (0.125in) dia x 3.175mm (0.125in) length, 99.95% (metals basis excluding Ta)
Thermo Scientific Chemicals

Niobium slug, 3.175mm (0.125in) dia x 3.175mm (0.125in) length, 99.95% (metals basis excluding Ta)

For vapor deposition | CAS: 7440-03-1 | Nb | 92.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 042845.09
also known as 042845-09
Price (USD)
58.65
Online Exclusive
64.50
Save 5.85 (9%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
58.65
Online Exclusive
64.50
Save 5.85 (9%)
Each
Chemical Identifiers
CAS7440-03-1
IUPAC Nameniobium
Molecular FormulaNb
InChI KeyGUCVJGMIXFAOAE-UHFFFAOYSA-N
SMILES[Nb]
View more
SpecificationsSpecification SheetSpecification Sheet
Trace Metalmetal impurities 0.05%
For vapor deposition

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only