Niobium slug, 6.35mm (0.25in) dia x 6.35mm (0.25in) length, 99.95% (metals basis excluding Ta)
Niobium slug, 6.35mm (0.25in) dia x 6.35mm (0.25in) length, 99.95% (metals basis excluding Ta)
Niobium slug, 6.35mm (0.25in) dia x 6.35mm (0.25in) length, 99.95% (metals basis excluding Ta)
Thermo Scientific Chemicals

Niobium slug, 6.35mm (0.25in) dia x 6.35mm (0.25in) length, 99.95% (metals basis excluding Ta)

CAS: 7440-03-1 | Nb | 92.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 042847.22
Price (USD)
220.00
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
220.00
Each
Chemical Identifiers
CAS7440-03-1
IUPAC Nameniobium
Molecular FormulaNb
InChI KeyGUCVJGMIXFAOAE-UHFFFAOYSA-N
SMILES[Nb]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Tantalum: Report Value
FormSlug
Total Metal Impurities≤0.05%, excluding Tantalum
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only