Rhodium sponge, -20 mesh, 99.9% (metals basis)
Rhodium sponge, -20 mesh, 99.9% (metals basis)
Rhodium sponge, -20 mesh, 99.9% (metals basis)
Thermo Scientific Chemicals

Rhodium sponge, -20 mesh, 99.9% (metals basis)

CAS: 7440-16-6 | Rh | 102.91 g/mol
Have Questions?
Catalog NumberQuantity
042866.03
also known as 042866-03
1 g
Catalog number 042866.03
also known as 042866-03
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Specifications
Assay Percent Notes(metals basis)
CAS7440-16-6
ChEBICHEBI:33359
Chemical Name or MaterialRhodium sponge
DOT InformationTransport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only