Chemical Name or MaterialPlatinum 10 wt% Rhodium wire
MDL NumberMFCD00198186
Name Note0.203mm (0.008 in.) dia., ISA Type S Standard Grade Thermocouple
OdorOdorless
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.