Platinum 6 wt% Rhodium wire, 0.5mm (0.02in) dia, ISA Type B Standard Grade Thermocouple
Thermo Scientific Chemicals

Platinum 6 wt% Rhodium wire, 0.5mm (0.02in) dia, ISA Type B Standard Grade Thermocouple

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 cm
100 cm
Catalog number 042914.BU
also known as 042914-BU
Price (USD)
-
Quantity:
25 cm
Request bulk or custom format
Specifications
Avg. Mol. Wt. or Mol. Wt. RangePt:Rh; 94:6 wt%
Chemical Name or MaterialPlatinum 6 wt% Rhodium wire
MDL NumberMFCD00198186
Name Note0.5mm (0.02 in.) dia., ISA Type B Standard Grade Thermocouple
OdorOdorless
View more
Platinum rhodium wire is used as a catalyst in preparation of nitric acid and in rayon spinnerets. It is used as a device for measuring high temperatures. It serves as a catalyst in the oxidation of the ammonia by atmospheric oxygen to produce nitric oxide, through Haber process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only