Lead granules, 99.999% (metals basis)
Lead granules, 99.999% (metals basis)
Lead granules, 99.999% (metals basis)
Thermo Scientific Chemicals

Lead granules, 99.999% (metals basis)

CAS: 7439-92-1 | Pb | 207.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 042928.30
also known as 042928-30
Price (USD)
441.65
Online Exclusive
490.00
Save 48.35 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
441.65
Online Exclusive
490.00
Save 48.35 (10%)
Each
Specifications
AppearanceGray granules
Assay Percent Range99.999% (Metals basis)
CAS7439-92-1
ChEBICHEBI:27889
Chemical Name or MaterialLead
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only