Tetrakis(dimethylamino)titanium(IV), 99.9% (metals basis)
Tetrakis(dimethylamino)titanium(IV), 99.9% (metals basis)
Tetrakis(dimethylamino)titanium(IV), 99.9% (metals basis)
Thermo Scientific Chemicals

Tetrakis(dimethylamino)titanium(IV), 99.9% (metals basis)

CAS: 3275-24-9 | C8H24N4Ti | 224.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 042932.06
also known as 042932-06
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Specifications
Assay Percent Notes(metals basis)
CAS3275-24-9
Chemical Name or MaterialTetrakis (dimethylamino)titanium (IV)
ColorBrown-Yellow
DOT InformationHazard Class: 4.3; Packaging Group: I
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only