Tantalum wire, 1.5mm (0.06in) dia, annealed, 99.95% (metals basis)
Tantalum wire, 1.5mm (0.06in) dia, annealed, 99.95% (metals basis)
Tantalum wire, 1.5mm (0.06in) dia, annealed, 99.95% (metals basis)
Thermo Scientific Chemicals

Tantalum wire, 1.5mm (0.06in) dia, annealed, 99.95% (metals basis)

CAS: 7440-25-7 | Ta | 180.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
20 cm
100 cm
500 cm
Catalog number 042933.BY
also known as 042933-BY
Price (USD)
275.65
Online Exclusive
306.00
Save 30.35 (10%)
Each
Quantity:
100 cm
Request bulk or custom format
Price (USD)
275.65
Online Exclusive
306.00
Save 30.35 (10%)
Each
Specifications
Assay Percent Notes(metals basis)
CAS7440-25-7
ChEBICHEBI:33348
Chemical Name or MaterialTantalum wire
EINECS Number231-135-5
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only