Boron trifluoride-dimethyl sulfide complex, purified, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Boron trifluoride-dimethyl sulfide complex, purified, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Boron trifluoride-dimethyl sulfide complex, purified, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Boron trifluoride-dimethyl sulfide complex, purified, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
042970.AE
also known as 042970-AE
100 mL
Catalog number 042970.AE
also known as 042970-AE
Price (USD)
318.65
Online Exclusive
354.00
Save 35.35 (10%)
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
318.65
Online Exclusive
354.00
Save 35.35 (10%)
Each
Chemical Identifiers
CAS353-43-5
IUPAC Name(methylsulfanyl)methane; trifluoroborane
Molecular FormulaC2H6BF3S
InChI KeyBRWZPVRDOUWXKE-UHFFFAOYSA-N
SMILESCSC.FB(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (Aqueous acid-base Titration)94-106%
Appearance (Color)Colorless to yellow to orange
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under Argon
RUO – Research Use Only