Gold Germanium slug, 3.175mm (0.125 in.) dia. x≈3.175mm (0.125 in.) length, Premion™, 99.998% (metals basis)
Thermo Scientific Chemicals

Gold Germanium slug, 3.175mm (0.125 in.) dia. x≈3.175mm (0.125 in.) length, Premion™, 99.998% (metals basis)

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 Pc.
25 Pc.
Catalog number 042983.KJ
also known as 042983-KJ
Price (USD)
539.00
Each
Quantity:
5 Pc.
Request bulk or custom format
Price (USD)
539.00
Each
Specifications
Assay Percent Notes(metals basis)
Avg. Mol. Wt. or Mol. Wt. RangeAu:Ge; 88:12 wt%
Chemical Name or MaterialGold Germanium slug
Density0.37 g/cm3
MDL NumberMFCD02091706
View more
Gold Germanium slug is used in scientific research and development activities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only