Silver slug, 3.175mm (0.125in) dia x 6.35mm (0.25in) length, Premion™, 99.999% (metals basis)
Silver slug, 3.175mm (0.125in) dia x 6.35mm (0.25in) length, Premion™, 99.999% (metals basis)
Silver slug, 3.175mm (0.125in) dia x 6.35mm (0.25in) length, Premion™, 99.999% (metals basis)
Thermo Scientific Chemicals

Silver slug, 3.175mm (0.125in) dia x 6.35mm (0.25in) length, Premion™, 99.999% (metals basis)

Form: Approx. 0.53g/slug | CAS: 7440-22-4 | Ag | 107.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 042985.09
also known as 042985-09
Price (USD)
132.00
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
132.00
Each
Chemical Identifiers
CAS7440-22-4
IUPAC Namesilver
Molecular FormulaAg
InChI KeyBQCADISMDOOEFD-UHFFFAOYSA-N
SMILES[Ag]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silver
Assay from Supplier's CofA≥99.999% (metals basis)
FormSlug, 3.175mm (0.125in) diameter.
Length6.35mm (0.25in)
Silver slug is used in thermal evaporation and electrodes (Schottky diode and fast diode).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only