Cesium, 99.8% (metals basis)
Cesium, 99.8% (metals basis)
Cesium, 99.8% (metals basis)
Thermo Scientific Chemicals

Cesium, 99.8% (metals basis)

CAS: 7440-46-2 | Cs | 132.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
50 g
Catalog number 043005.06
also known as 043005-06
Price (USD)
243.65
Online Exclusive
270.00
Save 26.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
243.65
Online Exclusive
270.00
Save 26.35 (10%)
Each
Chemical Identifiers
CAS7440-46-2
IUPAC Namecaesium
Molecular FormulaCs
InChI KeyTVFDJXOCXUVLDH-UHFFFAOYSA-N
SMILES[Cs]
View more
SpecificationsSpecification SheetSpecification Sheet
FormSolid
Appearance (Color)Yellow
Total Metal Impurities≤0.2% (2000 ppm)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only