Platinum Iridium wire, 0.127mm (0.005in) dia, annealed, 99.9% (metals basis excluding precious metals)
Thermo Scientific Chemicals

Platinum Iridium wire, 0.127mm (0.005in) dia, annealed, 99.9% (metals basis excluding precious metals)

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 cm
50 cm
Catalog number 043019.BS
also known as 043019-BS
Price (USD)
-
Quantity:
10 cm
Request bulk or custom format
Specifications
Assay Percent Notes(metals basis excluding precious metals)
Avg. Mol. Wt. or Mol. Wt. RangePt:Ir; 90:10 wt%
Chemical Name or MaterialPlatinum Iridium wire
MDL NumberMFCD02091683
Name Note0.127mm (0.005 in.) dia., Annealed, Temper: soft
View more
Vacuum evaporation deposition, scanning and tunneling microscopy, corrosion resistant electrodes.Platinum-iridium wire is used in platinum jewellery, particularly watch cases. They have been used for spinnerets in the manufacture of synthetic fibres, in metrology and alloy is fabrication of metal microelectrodes. It is used in electrical stimulation of nervous tissue and electrophysiological recordings. Pt/Ir alloy has been used in mechanical and electrochemical devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Vacuum evaporation deposition, scanning and tunneling microscopy, corrosion resistant electrodes
RUO – Research Use Only