Iron(III) isopropoxide, 2.5% w/v in isopropanol
Iron(III) isopropoxide, 2.5% w/v in isopropanol
Iron(III) isopropoxide, 2.5% w/v in isopropanol
Thermo Scientific Chemicals

Iron(III) isopropoxide, 2.5% w/v in isopropanol

CAS: 14995-22-3 | C9H21FeO3 | 233.109 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
Catalog number 043024.AA
also known as 043024-AA
Price (USD)
43.40
Each
Quantity:
5 mL
Request bulk or custom format
Price (USD)
43.40
Each
Specifications
CAS14995-22-3
Chemical Name or MaterialIron (III) isopropoxide
ColorRed to Brown
Concentration or Composition (by Analyte or Components)2.5% w/v in isopropanol
DOT InformationTransport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL
View more

Iron(III) isopropoxide is used in nano-materials & thin films, engineering & environmental, catalysis & synthesis and energy conversion & storage.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
RUO – Research Use Only