Titanium sputtering target, 76.2mm (3.0in) dia x 6.35mm (0.250in) thick, 99.995% (metals basis)
Titanium sputtering target, 76.2mm (3.0in) dia x 6.35mm (0.250in) thick, 99.995% (metals basis)
Titanium sputtering target, 76.2mm (3.0in) dia x 6.35mm (0.250in) thick, 99.995% (metals basis)
Thermo Scientific Chemicals

Titanium sputtering target, 76.2mm (3.0in) dia x 6.35mm (0.250in) thick, 99.995% (metals basis)

CAS: 7440-32-6 | Ti | 47.87 g/mol
Have Questions?
Catalog NumberQuantity
043045.KS1 Ea.
Catalog number 043045.KS
Price (USD)
341.65
Online Exclusive
379.00
Save 37.35 (10%)
Each
Quantity:
1 Ea.
Request bulk or custom format
Price (USD)
341.65
Online Exclusive
379.00
Save 37.35 (10%)
Each
Chemical Identifiers
CAS7440-32-6
IUPAC Nametitanium
Molecular FormulaTi
InChI KeyRTAQQCXQSZGOHL-UHFFFAOYSA-N
SMILES[Ti]
View more
SpecificationsSpecification SheetSpecification Sheet
FormSputtering target, 76.2mm (3.0in) diameter x 6.35mm (0.250in) thick.
Appearance (Color)Gray
Assay from Supplier's CofA≥99.995% (metals basis)
As intermediates and as salt. Used in coating, vesicant and powder metallurgy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only