Niobium rod, 19mm (0.75in) dia, hard, 99.8% (metals basis)
Niobium rod, 19mm (0.75in) dia, hard, 99.8% (metals basis)
Niobium rod, 19mm (0.75in) dia, hard, 99.8% (metals basis)
Thermo Scientific Chemicals

Niobium rod, 19mm (0.75in) dia, hard, 99.8% (metals basis)

CAS: 7440-03-1 | Nb | 92.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 cm
10 cm
Catalog number 043080.BP
also known as 043080-BP
Price (USD)
101.65
Online Exclusive
113.00
Save 11.35 (10%)
Each
Quantity:
2 cm
Request bulk or custom format
Price (USD)
101.65
Online Exclusive
113.00
Save 11.35 (10%)
Each
Chemical Identifiers
CAS7440-03-1
IUPAC Nameniobium
Molecular FormulaNb
InChI KeyGUCVJGMIXFAOAE-UHFFFAOYSA-N
SMILES[Nb]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥99.8% (metals basis)
FormRod, 19mm (0.75in) diameter, hardened.
Appearance (Color)Silvery
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only