Cesium acetate, 99% (metals basis)
Cesium acetate, 99% (metals basis)
Cesium acetate, 99% (metals basis)
Thermo Scientific Chemicals

Cesium acetate, 99% (metals basis)

CAS: 3396-11-0 | C2H3CsO2 | 191.949 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 043095.18
also known as 043095-18
Price (USD)
79.65
Special offer
Online exclusive
Ends: 28-Dec-2026
93.60
Save 13.95
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
79.65
Special offer
Online exclusive
Ends: 28-Dec-2026
93.60
Save 13.95
Each
Chemical Identifiers
CAS3396-11-0
IUPAC Namecaesium(1+) acetate
Molecular FormulaC2H3CsO2
InChI KeyZOAIGCHJWKDIPJ-UHFFFAOYSA-M
SMILES[Cs+].CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLump/Powder
Appearance (Color)White to light brown
Total Metal Impurities1% max.
Cesium acetate is used in organic synthesis especially in Perkin reaction to prepare unsaturated cinnamic-type acids. It is used in petroleum drilling fluids as an alternative to cesium formate. In organometallic chemistry, it is an additive for metal-ctalyzed coupling reactions. It is also used in the inversion of stereo chemistry of secondary alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only