Carbon, activated, -4+8 mesh
Carbon, activated, -4+8 mesh
Carbon, activated, -4+8 mesh
Thermo Scientific Chemicals

Carbon, activated, -4+8 mesh

CAS: 1333-86-4 | C | 12.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2 kg
Catalog number 043118.36
Price (USD)
56.65
Special offer
Online exclusive
Ends: 30-Jun-2026
66.90
Save 10.25 (15%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
56.65
Special offer
Online exclusive
Ends: 30-Jun-2026
66.90
Save 10.25 (15%)
Each
Specifications
CAS1333-86-4
Chemical Name or MaterialCarbon
EINECS Number231-153-3
Health Hazard 1H500
InChI KeyOKTJSMMVPCPJKN-UHFFFAOYSA-N
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only