Zirconium, Oil based standard solution, Specpure™ Zr 1000μg/g
Thermo Scientific Chemicals

Zirconium, Oil based standard solution, Specpure™ Zr 1000μg/g

 
Have Questions?
Catalog NumberQuantity
043127.18
also known as 043127-18
50 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 043127.18
also known as 043127-18
Price (USD)
-
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialZirconium, Oil based standard solution
MDL NumberMFCD00011303
Name NoteZr 1000μg/g
Physical FormLiquid
Quantity50 g
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only