Dionex™ IonPac™ CS11 IC Columns
Actual product may vary
Dionex™ IonPac™ CS11 IC Columns
Dionex™ IonPac™ CS11 IC Columns
Thermo Scientific™

Dionex™ IonPac™ CS11 IC Columns

Green features
Choose this low-capacity, sulfonate-functionalized, cation-exchange column for isocratic separation of inorganic cations using HCl with DAP eluents.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Column Type
04312850 mmGuard Column
043127250 mmAnalytical Column
Catalog number 043128
Price (MXN)
-
Length (Metric):
50 mm
Column Type:
Guard Column
Choose Thermo Scientific™ Dionex™ IonPac™CS11 Analytical and Guard Columns for isocratic separation of inorganic cations using hydrochloric acid (HCl) with diaminopropionic (DAP) acid eluents. This column has selectivity similar to the IonPac CS10 column, with improved sensitivity and lower eluent consumption due to its smaller 2mm I.D. format.
The Dionex IonPac CS11column can be used to determine common inorganic cations and aliphatic and aromatic amines in diverse sample matrices, however, advances in cation-exchange column technology have produced better column choices for these separations.

Recommended Cation-Exchange Columns by Use

Since the introduction of the Dionex IonPac CS11 column, Dionex has introduced several new cation-exchange columns with advanced features. For example, most modern Dionex IonPac Cation Exchange columns use simple methanesulfonic acid eluents rather than DAP-HCl eluents. These columns and their recommended uses are listed below.
  • The Dionex IonPac CS12A column improves performance for fast analysis of inorganic cations and ammonium in diverse matrices.
  • The Dionex IonPac CS17 column is recommended for hydrophobic and polyvalent amines.
  • The Dionex IonPac CS19 and CS19-4μm columns are recommended for polar amines including alkanolamines and methylamines and also moderately hydrophobic amines and polyvalent amines.
  • Compatible with organic solvents, to enhance analyte solubility, modify column selectivity, or allow effective column cleanup
Formats Available for Application Flexibility
  • 2x250mm analytical column
  • 2x50mm guard column
  • Longer 250mm columns facilitate higher resolution separations of more complex sample matrices
  • Smaller i.d. columns provide higher mass sensitivity, eluent and reagent conservation and lower operating costs
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionCG11; 2 x 50 mm
For Use With (Application)Cation Exchange
For Use With (Equipment)IonPac CS11 Analytical Column
Green FeaturesFSC
HydrophobicityMedium
Max. Pressure4000 psi (275 bar)
Particle Size8 μm
QuantityEach 1
Solvent CompatibilitypH 0 to 14, 0 to 100% HPLC Solvents (i.e. Methanol, Acetonitrile)
Stationary PhaseIonPac CG11
Column TypeGuard Column
Diameter (Metric)2 mm
FormatMicrobore
Length (Metric)50 mm
Unit SizeEach 1