Scandium, Oil based standard solution, Specpure™ Sc 1000μg/g
Thermo Scientific Chemicals

Scandium, Oil based standard solution, Specpure™ Sc 1000μg/g

 
Have Questions?
Catalog NumberQuantity
043129.18
also known as 043129-18
50 g
Catalog number 043129.18
also known as 043129-18
Price (USD)
115.65
Online Exclusive
128.00
Save 12.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
115.65
Online Exclusive
128.00
Save 12.35 (10%)
Each
Specifications
Chemical Name or MaterialScandium, Oil based standard solution
MDL NumberMFCD00016323
Name NoteSc 1000μg/g
Physical FormLiquid
Quantity50 g
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only