Magnesium oxide, 2 part ceramic adhesive paste
Magnesium oxide, 2 part ceramic adhesive paste
Magnesium oxide, 2 part ceramic adhesive paste
Thermo Scientific Chemicals

Magnesium oxide, 2 part ceramic adhesive paste

High Temperature Adhesive For Bonding Ceramic To Metal
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Catalog number 043135.36
also known as 043135-36
Price (USD)
161.65
Online Exclusive
179.00
Save 17.35 (10%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
161.65
Online Exclusive
179.00
Save 17.35 (10%)
Each
Specifications
Chemical Name or MaterialMagnesium oxide, 2 part ceramic adhesive paste
Health Hazard 1H314-H335-H303: Causes severe skin burns and eye damage. May cause respiratory irritation. May be harmful if swallowed.
MDL NumberMFCD00011109
Name NoteMix ratio (w/w): 1.5:1 powder to liquid. Cure conditions: 1-4 hours at room temperature, 2 hours at 94°C
Recommended StorageAmbient temperatures
View more
High temperature adhesive for bonding ceramic to metal

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Mix ratio (w/w): 1.5:1 powder to liquid. Cure conditions: 1-4 hours at room temperature, 2 hours at 94°C. Components: Magnesium oxide, 2 part ceramic adhesive paste (L) and Magnesium oxide, 2 part ceramic adhesive paste (P)
RUO – Research Use Only