Aluminum nitrate nonahydrate, low mercury, Puratronic™, 99.999% (metals basis)
Aluminum nitrate nonahydrate, low mercury, Puratronic™, 99.999% (metals basis)
Aluminum nitrate nonahydrate, low mercury, Puratronic™, 99.999% (metals basis)
Thermo Scientific Chemicals

Aluminum nitrate nonahydrate, low mercury, Puratronic™, 99.999% (metals basis)

CAS: 7784-27-2 | AlH18N3O18 | 375.129 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 043145.14
also known as 043145-14
Price (USD)
142.00
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
142.00
Each
Chemical Identifiers
CAS7784-27-2
IUPAC Namealuminium(3+) nonahydrate trinitrate
Molecular FormulaAlH18N3O18
InChI KeySWCIQHXIXUMHKA-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.O.O.O.[Al+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (special)Mercury (Hg): ≤1 ppm
Total Metal Impurities≤0.001%
Aluminum nitrate is a strong oxidizing agent. It is used in tanning leather, antiperspirants, corrosion inhibitors, extraction of uranium, petroleum refining, and as a nitrating agent. The nonahydrate and other hydrated aluminum nitrates have many applications. These salts are used to produce alumina for preparation of insulating papers, in cathode ray tube heating elements, and on transformer core laminates. The hydrated salts are also used for the extraction of actinide elements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only