Thermo Scientific Chemicals

Aluminum nitrate nonahydrate, low mercury, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 043145.22
100 g, Each
Thermo Scientific Chemicals

Aluminum nitrate nonahydrate, low mercury, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 043145.22
100 g, Each
Quantity
Catalog number: 043145.22
also known as 043145-22
Price (USD)
Price: 349.00
Online price: 296.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7784-27-2
IUPAC Name
aluminium(3+) nonahydrate trinitrate
Molecular Formula
AlH18N3O18
InChI Key
SWCIQHXIXUMHKA-UHFFFAOYSA-N
SMILES
O.O.O.O.O.O.O.O.O.[Al+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Assay (special)
Mercury (Hg): ≤1 ppm
Total Metal Impurities
≤0.001%

Description

Aluminum nitrate is a strong oxidizing agent. It is used in tanning leather, antiperspirants, corrosion inhibitors, extraction of uranium, petroleum refining, and as a nitrating agent. The nonahydrate and other hydrated aluminum nitrates have many applications. These salts are used to produce alumina for preparation of insulating papers, in cathode ray tube heating elements, and on transformer core laminates. The hydrated salts are also used for the extraction of actinide elements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text