4-Methyl-2-pentanone, HPLC Grade, 99+%
4-Methyl-2-pentanone, HPLC Grade, 99+%
4-Methyl-2-pentanone, HPLC Grade, 99+%
Thermo Scientific Chemicals

4-Methyl-2-pentanone, HPLC Grade, 99+%

Maximum level of impurities: Evaporation residue 5ppm, H2O 0.1%, Color (APHA) 10 | CAS: 108-10-1 | C6H12O | 100.161 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 L
4 x 1 L
4 x 4 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 043170.K2
also known as 043170-K2
Price (USD)
112.00
Each
Quantity:
1 L
Request bulk or custom format
Price (USD)
112.00
Each
Chemical Identifiers
CAS108-10-1
IUPAC Name4-methylpentan-2-one
Molecular FormulaC6H12O
InChI KeyNTIZESTWPVYFNL-UHFFFAOYSA-N
SMILESCC(C)CC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Residue on Evaporation5ppm
Residual water0.1%
Color (APHA)10
4-Methyl-2-pentanone is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only