Lithium hydroxide monohydrate, ACS, 98% min
Lithium hydroxide monohydrate, ACS, 98% min
Lithium hydroxide monohydrate, ACS, 98% min
Lithium hydroxide monohydrate, ACS, 98% min
Thermo Scientific Chemicals

Lithium hydroxide monohydrate, ACS, 98% min

CAS: 1310-66-3 | H3LiO2 | 41.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2 kg
Catalog number 043171.22
also known as 043171-22
Price (USD)
54.10
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
54.10
Each
Chemical Identifiers
CAS1310-66-3
IUPAC Namelithium(1+) hydrate hydroxide
Molecular FormulaH3LiO2
InChI KeyGLXDVVHUTZTUQK-UHFFFAOYSA-M
SMILES[Li+].O.[OH-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystalline powder or crystals or solid
Assay (Aqueous acid-base Titration)≥98.0%
Chloride content≤0.01%
Iron content≤0.002%
View more

Lithium chloride is useful for the production of lithium metal and for the generation of Mn(0) species which can be used in free radical cyclizations. It can serve as a flame colorant to generate dark red flames, a brazing flux for aluminum in automobiles, a hygrometer and a desiccant for drying air streams. On exposure to air, it becomes a solution with the concentration directly related to relative humidity of the atmosphere and hence serves as a relative humidity standard in calibrating hygrometers. Apart from being a source of chloride, it serves as an additive in the Stille reaction in organic synthesis and to precipitate RNA from cellular extracts. Being biologically significant, it finds applications in a wide variety of assays to study cell-fate and neurobiology. Lithium chloride has been found to inhibit virus infection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only