N,N'-Bis(salicylidene)ethylenediaminecobalt(II), 96%
N,N'-Bis(salicylidene)ethylenediaminecobalt(II), 96%
N,N'-Bis(salicylidene)ethylenediaminecobalt(II), 96%
Thermo Scientific Chemicals

N,N'-Bis(salicylidene)ethylenediaminecobalt(II), 96%

CAS: 14167-18-1 | C16H14CoN2O2 | 325.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
100 g
Catalog number 043174.06
also known as 043174-06
Price (USD)
43.65
Special offer
Online exclusive
Ends: 30-Jun-2026
50.70
Save 7.05 (14%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
43.65
Special offer
Online exclusive
Ends: 30-Jun-2026
50.70
Save 7.05 (14%)
Each
Chemical Identifiers
CAS14167-18-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange to red to brown
Purity≥95.0%
FormPowder or crystals or crystalline powder
Salcomine catalyzes the oxidation of 2,6-disubstituted phenols by dioxygen. Experimental inhibitor of human cytμloviris proteinase activity. Metal complex dyes used as redox catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only