Bis(cyclopentadienyl)chromium, sublimed, 97+%
Bis(cyclopentadienyl)chromium, sublimed, 97+%
Bis(cyclopentadienyl)chromium, sublimed, 97+%
Thermo Scientific Chemicals

Bis(cyclopentadienyl)chromium, sublimed, 97+%

CAS: 1271-24-5 | C10H10Cr | 182.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 043177.06
also known as 043177-06
Price (USD)
478.65
Online Exclusive
532.00
Save 53.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
478.65
Online Exclusive
532.00
Save 53.35 (10%)
Each
Chemical Identifiers
CAS1271-24-5
IUPAC NameBis(cyclopentadienyl)chromium
Molecular FormulaC10H10Cr
InChI KeyTYYBBNOTQFVVKN-UHFFFAOYSA-N
SMILES[Cr++].[CH-]1C=CC=C1.[CH-]1C=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Red to purple to black
FormCrystals or crystalline powder
Assay from Supplier's CofA≥97.0%
Provides catalyst for ethylene polymerization. A precursor to other chromium(II) compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
RUO – Research Use Only