Cerium(III) nitrate hydrate, REacton™, 99.998% (metals basis)
Thermo Scientific Chemicals

Cerium(III) nitrate hydrate, REacton™, 99.998% (metals basis)

Ce(NO3)3·xH2O (x ≈ 6-7)
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 043181.14
also known as 043181-14
Price (USD)
151.65
Online exclusive
168.00
Save 16.35
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
151.65
Online exclusive
168.00
Save 16.35
Each
Specifications
AppearanceWhite crystalline
Assay Percent Range99.998%
Chemical Name or MaterialCerium(III) nitrate hydrate
ColorWhite
DOT InformationDOT Class: 5.1, Packaging Group: III
View more
Cerium(III) nitrate hydrate is used as an analytical reagent. It acts as a catalyst in petrochemical industry and in the pharmaceutical industry. Further, it is a chemical that helps burn tissue recover in vitro.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only