Thermo Scientific Chemicals

Óxido de magnesio, 99,95% (base metálica), Thermo Scientific Chemicals

Número de catálogo: 043196.22
100 g, Each
Thermo Scientific Chemicals

Óxido de magnesio, 99,95% (base metálica), Thermo Scientific Chemicals

Número de catálogo: 043196.22
100 g, Each
Cantidad
Número de catálogo: 043196.22
también denominado 043196-22
Precio (EUR)
Cantidad
-

Identificadores químicos

CAS
1309-48-4
IUPAC Name
oxomagnesium
Molecular Formula
MgO
InChI Key
CPLXHLVBOLITMK-UHFFFAOYSA-N
SMILES
O=[Mg]
Total Metal Impurities
0.05% max.

Descripción

Magnesium oxide is used as a laboratory reagent, and a good reference white color for colorimetry. Magneisum oxide microspheres is used for enrichment method of polycyclic aromatic hydrocarbons. It is also used in groundwater studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text