Benzyl acrylate, 98%, stab. with ca 150ppm 4-methoxyphenol
Benzyl acrylate, 98%, stab. with ca 150ppm 4-methoxyphenol
Benzyl acrylate, 98%, stab. with ca 150ppm 4-methoxyphenol
Thermo Scientific Chemicals

Benzyl acrylate, 98%, stab. with ca 150ppm 4-methoxyphenol

Preparation of high refractive index polyacrylates | CAS: 2495-35-4 | C10H10O2 | 162.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number 043204.09
also known as 043204-09
Price (USD)
45.70
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
45.70
Each
Chemical Identifiers
CAS2495-35-4
IUPAC Namebenzyl prop-2-enoate
Molecular FormulaC10H10O2
InChI KeyGCTPMLUUWLLESL-UHFFFAOYSA-N
SMILESC=CC(=O)OCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥96.0%
Preparation of high refractive index polyacrylates.Benzyl acrylate is used in the preparation of heptanoic acid benzyl ester. It is used to prepare polybenzylacrylate using azobisisobutyronitrile as initiator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only