Poly(methyl-3,3,3-trifluoropropylsiloxane), M.W. 2,400
Poly(methyl-3,3,3-trifluoropropylsiloxane), M.W. 2,400
Poly(methyl-3,3,3-trifluoropropylsiloxane), M.W. 2,400
Thermo Scientific Chemicals

Poly(methyl-3,3,3-trifluoropropylsiloxane), M.W. 2,400

Fluorosilicone lubricant | CAS: 63148-56-1 | C4H7F3OSi | 156.179 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 043228.14
also known as 043228-14
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS63148-56-1
IUPAC Namemethyl(3,3,3-trifluoropropyl)silanone
Molecular FormulaC4H7F3OSi
InChI KeyCUMCLKHSZZGALD-UHFFFAOYSA-N
SMILESC[Si](=O)CCC(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Viscosity270-385 cSt
Identification (FTIR)Conforms
Appearance (Color)Clear colorless
FormLiquid
Fluorosilicone lubricantPoly(methyl-3,3,3-trifluoropropylsiloxane) is used in the preparation of siliconized- glass fiber sheets, which acts as a support for protein-chemical analysis of electroblotted proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only